Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338223.1 | NP_001327485.1 | 4 - 5 | 776 | 111 | |
NM_001338224.1 | NP_001327484.1 | 4 - 5 | 763 | 111 | ||
NM_112497.3 | NP_188247.2 | 5 - 6 | 685 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |