Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338228.1 | NP_001319567.1 | 1 - 2 | 327 | 66 | |
NM_001338229.1 | NP_001325497.1 | 1 - 2 | 255 | 66 | ||
NM_001338230.1 | NP_001325498.1 | 1 - 2 | 195 | 66 | ||
NM_112503.5 | NP_188253.3 | 1 - 2 | 327 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |