Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338255.1 | NP_001326711.1 | 17 - 18 | 3354 | 70 | |
NM_001338256.1 | NP_001326710.1 | 17 - 18 | 3354 | 70 | ||
NM_001338257.1 | NP_001326709.1 | 17 - 18 | 3308 | 70 | ||
NM_112553.4 | NP_188302.2 | 17 - 18 | 3354 | 70 | ||
GenBank mRNA | AF411833.1 | - | 17 - 18 | 3161 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |