Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338374.1 | NP_001326749.1 | 2 - 3 | 973 | 128 | |
NM_112785.5 | NP_188529.2 | 2 - 3 | 994 | 128 | ||
GenBank mRNA | AB493620.1 | - | 2 - 3 | 507 | 128 | |
AF289052.1 | - | 2 - 3 | 818 | 128 | ||
AK230388.1 | - | 706 | 128 | |||
BT024731.1 | - | 2 - 3 | 507 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |