Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203004.1 | NP_001189933.1 | 14 - 15 | 1911 | 69 | |
NM_001338387.1 | NP_001325757.1 | 14 - 15 | 1892 | 69 | ||
NM_112808.4 | NP_188552.3 | 14 - 15 | 1892 | 69 | ||
GenBank mRNA | AB250666.1 | - | 14 - 15 | 1867 | 69 | |
DQ912973.1 | - | 14 - 15 | 1805 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |