Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATHXK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112895.3 | NP_188639.2 | 8 - 9 | 1690 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |