Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342374.1 | NP_001329346.1 | 6 - 7 | 1431 | 106 | |
NM_119746.4 | NP_195305.2 | 6 - 7 | 1431 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |