Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCD5;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342451.1 | NP_001328589.1 | 2 - 3 | 658 | 95 | |
NM_119926.5 | NP_195478.2 | 2 - 3 | 658 | 95 | ||
GenBank mRNA | AK221431.1 | - | 2 - 3 | 547 | 95 | |
BT011732.1 | - | 2 - 3 | 329 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |