Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119938.4 | NP_568033.1 | 5 - 6 | 1802 | 80 | |
GenBank mRNA | AK226847.1 | - | 5 - 6 | 1568 | 80 | |
AY072019.1 | - | 5 - 6 | 1543 | 80 | ||
BT001057.1 | - | 5 - 6 | 1455 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |