Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202724.2 | NP_001189653.1 | 40 - 41 | 6141 | 103 | |
NM_001336361.1 | NP_001318331.1 | 41 - 42 | 5853 | 103 | ||
NM_001336362.1 | NP_001323730.1 | 41 - 42 | 5607 | 103 | ||
NM_001336363.1 | NP_001323731.1 | 41 - 42 | 5778 | 103 | ||
NM_179847.3 | NP_850178.2 | 40 - 41 | 5781 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |