Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT2G34150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036397.3 | NP_001031474.1 | 7 - 8 | 2509 | 74 | |
NM_001336486.1 | NP_001324986.1 | 4 - 5 | 2143 | 74 | ||
NM_001336487.1 | NP_001324987.1 | 6 - 7 | 2186 | 74 | ||
NM_001336488.1 | NP_001324988.1 | 5 - 6 | 2182 | 74 | ||
NM_179894.2 | NP_850225.1 | 7 - 8 | 2535 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |