Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129360.4 | NP_565880.1 | 3 - 4 | 790 | 91 | |
NM_179960.1 | NP_850291.1 | 3 - 4 | 704 | 91 | ||
GenBank mRNA | AF056969.1 | - | 3 - 4 | 660 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |