Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129360.4 | NP_565880.1 | 8 - 9 | 1377 | 76 | |
NM_179960.1 | NP_850291.1 | 8 - 9 | 1291 | 76 | ||
GenBank mRNA | AF056969.1 | - | 8 - 9 | 1247 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |