Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123738.2 | NP_001117210.1 | 2 - 3 | 533 | 96 | |
NM_001331281.1 | NP_001322112.1 | 2 - 3 | 541 | 96 | ||
NM_001331282.1 | NP_001322113.1 | 2 - 3 | 508 | 96 | ||
NM_100038.4 | NP_563631.2 | 2 - 3 | 508 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |