Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123738.2 | NP_001117210.1 | 3 - 4 | 664 | 76 | |
NM_001331281.1 | NP_001322112.1 | 3 - 4 | 672 | 76 | ||
NM_001331282.1 | NP_001322113.1 | 3 - 4 | 639 | 76 | ||
NM_100038.4 | NP_563631.2 | 3 - 4 | 639 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |