Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160828.1 | NP_001154300.1 | 10 - 11 | 1602 | 128 | |
NM_001160829.1 | NP_001154301.1 | 10 - 11 | 1602 | 128 | ||
NM_001331317.1 | NP_001320318.1 | 9 - 10 | 1744 | 128 | ||
NM_100075.8 | NP_171697.3 | 10 - 11 | 1665 | 128 | ||
GenBank mRNA | AB290929.1 | - | 10 - 11 | 1514 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |