Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197959.1 | NP_001184888.1 | 20 - 21 | 1987 | 109 | |
NM_001331318.1 | NP_001320381.1 | 19 - 20 | 2291 | 109 | ||
NM_001331319.1 | NP_001320383.1 | 19 - 20 | 2266 | 109 | ||
NM_001331320.1 | NP_001320382.1 | 19 - 20 | 2260 | 109 | ||
NM_001331321.1 | NP_001320384.1 | 14 - 15 | 1554 | 109 | ||
NM_100081.5 | NP_563643.4 | 20 - 21 | 2387 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |