Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100148.3 | NP_171768.2 | 2 - 3 | 239 | 67 | |
GenBank mRNA | AB493426.1 | - | 1 - 2 | 104 | 67 | |
AK175906.1 | - | 2 - 3 | 239 | 67 | ||
AK175966.1 | - | 2 - 3 | 239 | 67 | ||
AK176854.1 | - | 2 - 3 | 207 | 67 | ||
AK221393.1 | - | 2 - 3 | 218 | 67 | ||
AY463615.1 | - | 1 - 2 | 104 | 67 | ||
BT010482.1 | - | 1 - 2 | 104 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |