Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100351.5 | NP_171965.1 | 1 - 2 | 140 | 80 | |
GenBank mRNA | AY099741.1 | - | 1 - 2 | 116 | 80 | |
BT003415.1 | - | 1 - 2 | 84 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |