Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT71C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100599.4 | NP_563784.2 | 1 | 1661 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |