Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100608.5 | NP_172214.5 | 1 - 2 | 183 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |