Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112950.3 | NP_188694.2 | 1 - 2 | 832 | 104 | |
GenBank mRNA | AK118605.1 | - | 1 - 2 | 747 | 104 | |
BT005952.1 | - | 1 - 2 | 600 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |