Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337111.1 | NP_001318427.1 | 5 - 6 | 1714 | 102 | |
NM_001337112.1 | NP_001324720.1 | 5 - 6 | 1744 | 102 | ||
NM_130064.5 | NP_182026.5 | 5 - 6 | 1767 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |