Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202819.1 | NP_001189748.1 | 4 - 5 | 941 | 69 | |
NM_130083.5 | NP_182045.1 | 4 - 5 | 794 | 69 | ||
GenBank mRNA | AF357529.1 | - | 4 - 5 | 543 | 69 | |
BT024623.1 | - | 4 - 5 | 488 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |