Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIMIN-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100812.3 | NP_563843.1 | 1 | 490 | 145 | |
GenBank mRNA | AF386947.1 | - | 1 | 396 | 145 | |
AY072486.1 | - | 1 | 330 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |