Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2A' | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100848.4 | NP_172447.1 | 5 - 6 | 713 | 76 | |
GenBank mRNA | AY059902.1 | - | 5 - 6 | 676 | 76 | |
BT000143.1 | - | 5 - 6 | 624 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |