Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100868.3 | NP_172465.1 | 1 - 2 | 528 | 90 | |
GenBank mRNA | BT023439.1 | - | 1 - 2 | 439 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |