Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331877.1 | NP_001322343.1 | 3 - 4 | 723 | 100 | |
NM_100875.1 | NP_172472.1 | 3 - 4 | 633 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |