Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQ4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100968.3 | NP_172562.2 | 20 - 21 | 3018 | 112 | |
GenBank mRNA | AY120761.1 | - | 20 - 21 | 2916 | 112 | |
BT010133.1 | - | 19 - 20 | 2712 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |