Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345215.1 | NP_001331990.1 | 2 - 3 | 530 | 133 | |
NM_125057.4 | NP_200485.1 | 4 - 5 | 459 | 133 | ||
GenBank mRNA | BT025320.1 | - | 4 - 5 | 297 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |