Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATLIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125098.4 | NP_568851.2 | 2 - 3 | 566 | 93 | |
GenBank mRNA | AF233527.1 | - | 2 - 3 | 486 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |