Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125192.4 | NP_200615.1 | 1 - 2 | 629 | 78 | |
GenBank mRNA | AY062789.1 | - | 1 - 2 | 422 | 78 | |
AY081612.1 | - | 1 - 2 | 369 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |