Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125241.5 | NP_200663.2 | 2 - 3 | 2624 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |