Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125258.5 | NP_200679.1 | 5 - 6 | 545 | 88 | |
GenBank mRNA | AF192490.1 | - | 5 - 6 | 503 | 88 | |
AY048227.1 | - | 5 - 6 | 494 | 88 | ||
AY086471.1 | - | 5 - 6 | 523 | 88 | ||
AY094017.1 | - | 5 - 6 | 480 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |