Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345330.1 | NP_001318835.1 | 5 - 6 | 1725 | 107 | |
NM_001345331.1 | NP_001332082.1 | 6 - 7 | 1170 | 107 | ||
NM_001345332.1 | NP_001332083.1 | 5 - 6 | 1122 | 107 | ||
NM_125284.4 | NP_200702.2 | 4 - 5 | 1200 | 107 | ||
GenBank mRNA | BT015092.1 | - | 4 - 5 | 871 | 107 | |
BT015906.1 | - | 4 - 5 | 798 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |