Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345382.1 | NP_001330990.1 | 2 - 3 | 246 | 65 | |
NM_001345383.1 | NP_001330991.1 | 1 - 2 | 150 | 65 | ||
NM_125395.3 | NP_200810.1 | 2 - 3 | 322 | 65 | ||
GenBank mRNA | BT015359.1 | - | 2 - 3 | 322 | 65 | |
BT015890.1 | - | 2 - 3 | 246 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |