Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345489.1 | NP_001318859.1 | 5 - 6 | 853 | 78 | |
NM_001345490.1 | NP_001332309.1 | 5 - 6 | 938 | 78 | ||
NM_001345491.1 | NP_001332310.1 | 5 - 6 | 913 | 78 | ||
NM_001345492.1 | NP_001332311.1 | 5 - 6 | 913 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |