Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203666.1 | NP_001190595.1 | 9 - 10 | 1037 | 87 | |
NM_125617.3 | NP_201029.1 | 9 - 10 | 1030 | 87 | ||
GenBank mRNA | FJ708810.1 | - | 9 - 10 | 733 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |