Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103854.3 | NP_175389.1 | 1 | 1061 | 97 | |
GenBank mRNA | BT022077.1 | - | 1 | 948 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |