Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103869.4 | NP_564555.1 | 2 - 3 | 624 | 106 | |
GenBank mRNA | AY057512.1 | - | 2 - 3 | 574 | 106 | |
AY090315.1 | - | 2 - 3 | 501 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |