Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIIIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103919.5 | NP_175453.2 | 2 - 3 | 774 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |