Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333445.1 | NP_001323177.1 | 1 - 2 | 304 | 122 | |
NM_103942.3 | NP_175475.2 | 1 - 2 | 202 | 122 | ||
GenBank mRNA | BT000883.1 | - | 1 - 2 | 202 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |