Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121001.2 | NP_568215.2 | 7 - 8 | 776 | 75 | |
GenBank mRNA | AY063833.1 | - | 7 - 8 | 776 | 75 | |
AY091309.1 | - | 7 - 8 | 717 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |