Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203340.2 | NP_001190269.1 | 3 - 4 | 415 | 66 | |
NM_121023.4 | NP_568219.1 | 3 - 4 | 399 | 66 | ||
GenBank mRNA | AF424566.1 | - | 3 - 4 | 319 | 66 | |
AF428323.1 | - | 3 - 4 | 337 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |