Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161232.2 | NP_001154704.2 | 11 - 12 | 1608 | 134 | |
NM_121141.5 | NP_196664.4 | 10 - 11 | 1536 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |