Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HY5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343175.1 | NP_001330553.1 | 1 - 2 | 299 | 90 | |
NM_121164.5 | NP_568246.1 | 1 - 2 | 369 | 90 | ||
GenBank mRNA | AB005295.1 | - | 1 - 2 | 287 | 90 | |
AB005456.1 | - | 1 - 2 | 287 | 90 | ||
AK229362.1 | - | 1 - 2 | 249 | 90 | ||
BT025519.1 | - | 1 - 2 | 96 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |