Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121190.3 | NP_196713.1 | 10 - 11 | 1398 | 83 | |
GenBank mRNA | AY050765.1 | - | 10 - 11 | 1308 | 83 | |
AY079310.1 | - | 10 - 11 | 1265 | 83 | ||
U15034.1 | - | 10 - 11 | 1308 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |