Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128176.3 | NP_180187.1 | 4 - 5 | 763 | 126 | |
GenBank mRNA | BT029734.1 | - | 4 - 5 | 642 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |