Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128190.3 | NP_180201.1 | 26 - 27 | 3340 | 65 | |
GenBank mRNA | AY035110.1 | - | 26 - 27 | 2649 | 65 | |
FJ708701.1 | - | 26 - 27 | 2573 | 65 | ||
U47029.1 | - | 26 - 27 | 2623 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |