Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336070.1 | NP_001324176.1 | 2 - 3 | 279 | 127 | |
NM_128200.5 | NP_565622.1 | 2 - 3 | 302 | 127 | ||
GenBank mRNA | AK226505.1 | - | 2 - 3 | 245 | 127 | |
AY086218.1 | - | 2 - 3 | 273 | 127 | ||
AY150419.1 | - | 1 - 2 | 155 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |